1,4-Diethynyl-2-fluorobenzene
Supplier: Apollo Scientific
Warning
PC49311-1G PC49311-5G PC49311-250MG
APOSPC49311-1GEA 58 EUR
APOSPC49311-1G APOSPC49311-5G APOSPC49311-250MG
1,4-Diethynyl-2-fluorobenzene
1,4-Diethynyl-2-fluorobenzene
Formula: C₁₀H₅F
MW: 144.145 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22580725
CAS Number: 137000-66-9