4-Bromo-2-fluorophenylacetylene
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-1-ethynyl-2-fluorobenzene
PC49319-1G PC49319-250MG
APOSPC49319-1GEA 58 EUR
APOSPC49319-1G APOSPC49319-250MG
4-Bromo-2-fluorophenylacetylene
4-Bromo-2-fluorophenylacetylene
Formula: C₈H₄BrF
MW: 199.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02683523
CAS Number: 302912-33-0