Methyl 2-iodo-4-(trifluoromethyl)benzoate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-iodo-α,α,α-trifluoro-p-toluate, 3-Iodo-4-(methoxycarbonyl)benzotrifluoride
PC49380-1G PC49380-5G PC49380-25G
APOSPC49380-1GEA 46.4 EUR
APOSPC49380-1G APOSPC49380-5G APOSPC49380-25G
Methyl 2-iodo-4-(trifluoromethyl)benzoate 97%
Methyl 2-iodo-4-(trifluoromethyl)benzoate
Formula: C₉H₆F₃IO₂
MW: 330.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399512
CAS Number: 1236303-09-5