2,2,2-Trifluoro-2'-(trifluoromethyl)acetophenone 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2,2-Trifluoro-1-(2-(trifluoromethyl)phenyl)ethanone, 2,2,2-Trifluoro-1-[2-(trifluoromethyl)phenyl]ethan-1-one, 2-(Trifluoroacetyl)benzotrifluoride
PC49384-1G PC49384-5G PC49384-25G
APOSPC49384-1GEA 101 EUR
APOSPC49384-1G APOSPC49384-5G APOSPC49384-25G
2,2,2-Trifluoro-2'-(trifluoromethyl)acetophenone 96%
2,2,2-Trifluoro-2'-(trifluoromethyl)acetophenone
Formula: C₉H₄F₆O
MW: 242.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07380778
CAS Number: 42452-42-6