6-Bromo-8-fluoroimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
PC49397-1G PC49397-250MG
APOSPC49397-1GEA 188 EUR
APOSPC49397-1G APOSPC49397-250MG
6-Bromo-8-fluoroimidazo[1,2-a]pyridine
6-Bromo-8-fluoroimidazo[1,2-a]pyridine
Formula: C₇H₄BrFN₂
MW: 215.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13193294
CAS Number: 474709-06-3