Ethyl 7-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
Warning
PC49401-1G PC49401-250MG
APOSPC49401-1GEA 429 EUR
APOSPC49401-1G APOSPC49401-250MG
Ethyl 7-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 7-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₉FN₂O₂
MW: 208.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11976204
CAS Number: 1260798-14-8