2-Fluoro-4-methyl-5-nitrobenzamide
Supplier: Apollo Scientific
|
Warning
|
PC49450-1G
PC49450-250MG
APOSPC49450-1GEA
986
EUR
APOSPC49450-1G
APOSPC49450-250MG
2-Fluoro-4-methyl-5-nitrobenzamide
2-Fluoro-4-methyl-5-nitrobenzamide
Formula:
C₈H₇FN₂O₃ MW: 198.151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28797494 CAS Number: 1622004-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...