2-Bromo-4,5-difluorobenzyl bromide
Supplier: Apollo Scientific
Danger
Synonyms: 1-Bromo-2-(bromomethyl)-4,5-difluorobenzene, α,2-Dibromo-4,5-difluorotoluene
PC49499-1G PC49499-5G PC49499-250MG
APOSPC49499-1GEA 58 EUR
APOSPC49499-1G APOSPC49499-5G APOSPC49499-250MG
2-Bromo-4,5-difluorobenzyl bromide
2-Bromo-4,5-difluorobenzyl bromide
Formula: C₇H₄Br₂F₂
MW: 285.911 g/mol
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD18909668
CAS Number: 647862-95-1
UN: 3261
ADR: 8 III