Tris[(trifluoromethyl)sulphonyl]methane 70% in aqueous solution
Supplier: Apollo Scientific
Danger
PC49610-1G PC49610-250MG
APOSPC49610-1GEA 915 EUR
APOSPC49610-1G APOSPC49610-250MG
Tris[(trifluoromethyl)sulphonyl]methane 70% in aqueous solution
Tris[(trifluoromethyl)sulphonyl]methane
Formula: C₄HF₉O₆S₃
MW: 412.228 g/mol
Boiling Pt: 220 °C
Density: 1.38 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD16876755
CAS Number: 60805-12-1
UN: 3265
ADR: 8,III