4-Hydroxy-2,3,5-trifluorobenzoic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4-Carboxy-2,3,6-trifluorophenol, 2,3,5-Trifluoro-4-hydroxybenzoic acid
PC49652-1G PC49652-250MG
APOSPC49652-1GEA 232 EUR
APOSPC49652-1G APOSPC49652-250MG
4-Hydroxy-2,3,5-trifluorobenzoic acid
4-Hydroxy-2,3,5-trifluorobenzoic acid
Formula: C₇H₃F₃O₃
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD08458460
CAS Number: 156839-10-0