4-Methoxy-2,3,5-trifluorobenzoic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4-Carboxy-2,3,6-trifluoroanisole, 2,3,5-Trifluoro-p-anisic acid
PC49653-1G PC49653-250MG
APOSPC49653-1GEA 71.2 EUR
APOSPC49653-1G APOSPC49653-250MG
4-Methoxy-2,3,5-trifluorobenzoic acid
4-Methoxy-2,3,5-trifluorobenzoic acid
Formula: C₈H₅F₃O₃
MW: 206.119 g/mol
Melting Pt: 141…144 °C
Storage Temperature: Ambient
MDL Number: MFCD06797838
CAS Number: 1003709-67-8