2,2,2-Trifluoroethanesulphonic acid 95%
Supplier: Apollo Scientific
Danger
PC49665-1G PC49665-5G PC49665-250MG
APOSPC49665-1GEA 306 EUR
APOSPC49665-1G APOSPC49665-5G APOSPC49665-250MG
2,2,2-Trifluoroethanesulphonic acid 95%
2,2,2-Trifluoroethanesulphonic acid
Formula: C₂H₃F₃O₃S
Boiling Pt: 108…110 °C (1.5 mm Hg)
Melting Pt: 50…52 °C
Storage Temperature: Ambient
MDL Number: MFCD28515421
CAS Number: 1827-97-0
UN: 3261
ADR: 8,III