Methyl 2-bromo-6-chloro-4-fluorobenzoate 95%
Supplier: Apollo Scientific
Warning
PC49997-1G PC49997-250MG
APOSPC49997-1GEA 219 EUR
APOSPC49997-1G APOSPC49997-250MG
Methyl 2-bromo-6-chloro-4-fluorobenzoate 95%
Methyl 2-bromo-6-chloro-4-fluorobenzoate
Formula: C₈H₅BrClFO₂
MW: 267.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28733926
CAS Number: 1695489-54-3