2-Fluoro-5-methoxyphenol
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluoro-3-hydroxyanisole
PC500161-1G PC500161-5G
APOSPC500161-1GEA 144 EUR
APOSPC500161-1G APOSPC500161-5G
2-Fluoro-5-methoxyphenol
2-Fluoro-5-methoxyphenol
Formula: C₇H₇FO₂
Melting Pt: 25…26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16998752
CAS Number: 117902-16-6