4-Fluoro-5-iodo-2-methylanisole
Supplier: Apollo Scientific
Warning
PC500306-1G PC500306-250MG PC500306-5G
APOSPC500306-1GEA 288 EUR
APOSPC500306-1G APOSPC500306-250MG APOSPC500306-5G
4-Fluoro-5-iodo-2-methylanisole
4-Fluoro-5-iodo-2-methylanisole
Formula: C₈H₈FIO
MW: 266.051 g/mol
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD19440267
CAS Number: 741716-19-8