2-Fluoro-4-methyl-5-nitroanisole 95%
Supplier: Apollo Scientific
Warning
PC500308-1G PC500308-250MG
APOSPC500308-1GEA 488 EUR
APOSPC500308-1G APOSPC500308-250MG
2-Fluoro-4-methyl-5-nitroanisole 95%
2-Fluoro-4-methyl-5-nitroanisole
Formula: C₈H₈FNO₃
MW: 185.152 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24368898
CAS Number: 3108-02-9