2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzaldehyde 95%
Supplier: Apollo Scientific
Warning
PC500314-1G PC500314-250MG
APOSPC500314-1GEA 69.6 EUR
APOSPC500314-1G APOSPC500314-250MG
2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzaldehyde 95%
2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27975486
CAS Number: 1823327-58-7