2-Chloro-6-fluoro-3-nitrotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-4-fluoro-3-methyl-1-nitrobenzene, 2-Chloro-4-fluoro-3-methylnitrobenzene
PC500364-1G PC500364-5G PC500364-25G
APOSPC500364-1GEA 21.9 EUR
APOSPC500364-1G APOSPC500364-5G APOSPC500364-25G
2-Chloro-6-fluoro-3-nitrotoluene
2-Chloro-6-fluoro-3-nitrotoluene
Formula: C₇H₅ClFNO₂
MW: 189.572 g/mol
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD21603723
CAS Number: 90292-62-9