4-Bromo-5-chloro-2-fluorobenzaldehyde 95%
Supplier: Apollo Scientific
Warning
PC500401-1G PC500401-250MG PC500401-5G
APOSPC500401-1GEA 127 EUR
APOSPC500401-1G APOSPC500401-250MG APOSPC500401-5G
4-Bromo-5-chloro-2-fluorobenzaldehyde 95%
4-Bromo-5-chloro-2-fluorobenzaldehyde
Formula: C₇H₃BrClFO
MW: 237.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26142710
CAS Number: 1603584-72-0