2-Fluoro-4-hydroxy-5-nitrobenzoic acid
Supplier: Apollo Scientific
Warning
PC500431-1G PC500431-250MG PC500431-5G
APOSPC500431-1GEA 105 EUR
APOSPC500431-1G APOSPC500431-250MG APOSPC500431-5G
2-Fluoro-4-hydroxy-5-nitrobenzoic acid
2-Fluoro-4-hydroxy-5-nitrobenzoic acid
Formula: C₇H₄FNO₅
MW: 201.109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26853061
CAS Number: 1565054-28-5