2,3-Difluoro-4-iodobenzonitrile
Supplier: Apollo Scientific
Warning
PC500461-1G PC500461-250MG PC500461-5G
APOSPC500461-1GEA 317 EUR
APOSPC500461-1G APOSPC500461-250MG APOSPC500461-5G
2,3-Difluoro-4-iodobenzonitrile
2,3-Difluoro-4-iodobenzonitrile
Formula: C₇H₂F₂IN
MW: 264.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28014018
CAS Number: 943830-91-9
UN: 3439
ADR: 6.1 III