3-Bromo-6-chloro-2-fluorobenzoic acid
Supplier: Apollo Scientific
Warning
PC500462-1G PC500462-25G PC500462-5G
APOSPC500462-1GEA 46.4 EUR
APOSPC500462-1G APOSPC500462-25G APOSPC500462-5G
3-Bromo-6-chloro-2-fluorobenzoic acid
3-Bromo-6-chloro-2-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110256
CAS Number: 702640-51-5