4-(Bromomethyl)-3-fluorobenzoic acid
Supplier: Apollo Scientific
Danger
Synonyms: α-Bromo-3-fluoro-p-toluic acid, 4-Carboxy-2-fluorobenzyl bromide
PC500463-1G PC500463-250MG PC500463-5G
APOSPC500463-1GEA 708 EUR
APOSPC500463-1G APOSPC500463-250MG APOSPC500463-5G
4-(Bromomethyl)-3-fluorobenzoic acid
4-(Bromomethyl)-3-fluorobenzoic acid
Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11179690
CAS Number: 361456-46-4
UN: 3261
ADR: 8 III