4-(Bromomethyl)-2-fluorobenzoic acid
Supplier: Apollo Scientific
Danger
Synonyms: α-Bromo-2-fluoro-p-toluic acid, 4-Carboxy-3-fluorobenzyl bromide
PC500465-1G PC500465-250MG PC500465-5G PC500465-10G PC500465-25G
APOSPC500465-1GEA 453 EUR
APOSPC500465-1G APOSPC500465-250MG APOSPC500465-5G APOSPC500465-10G APOSPC500465-25G
4-(Bromomethyl)-2-fluorobenzoic acid
4-(Bromomethyl)-2-fluorobenzoic acid
Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06208847
CAS Number: 477199-77-2
UN: 3261
ADR: 8 III