3-(Bromomethyl)-5-fluorobenzoic acid
Supplier: Apollo Scientific
Danger
Synonyms: α-Bromo-5-fluoro-m-toluic acid, 3-Carboxy-5-fluorobenzyl bromide
PC500474-1G PC500474-250MG PC500474-5G
APOSPC500474-1GEA 319 EUR
APOSPC500474-1G APOSPC500474-250MG APOSPC500474-5G
3-(Bromomethyl)-5-fluorobenzoic acid
3-(Bromomethyl)-5-fluorobenzoic acid
Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18378634
CAS Number: 1379351-68-4
UN: 3261
ADR: 8 III