5-(Bromomethyl)-2-fluorobenzoic acid
Supplier: Apollo Scientific
Danger
Synonyms: 3-Carboxy-4-fluorobenzyl bromide, α-Bromo-6-fluoro-m-toluic acid
PC500479-1G PC500479-250MG PC500479-5G
APOSPC500479-1GEA 328 EUR
APOSPC500479-1G APOSPC500479-250MG APOSPC500479-5G
5-(Bromomethyl)-2-fluorobenzoic acid
5-(Bromomethyl)-2-fluorobenzoic acid
Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06208849
CAS Number: 773100-76-8
UN: 3261
ADR: 8 III