3-Bromo-6-chloro-2-fluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC500489-1G PC500489-250MG
APOSPC500489-1GEA 306 EUR
APOSPC500489-1G APOSPC500489-250MG
3-Bromo-6-chloro-2-fluoroaniline 98%
3-Bromo-6-chloro-2-fluoroaniline
Formula: C₆H₄BrClFN
Storage Temperature: Ambient
MDL Number: MFCD27934056
CAS Number: 943830-81-7