6-Bromo-3-iodo-2-fluoronitrobenzene
Supplier: Apollo Scientific
Warning
PC500521-100MG PC500521-250MG
APOSPC500521-100MGEA 130 EUR
APOSPC500521-100MG APOSPC500521-250MG
6-Bromo-3-iodo-2-fluoronitrobenzene
6-Bromo-3-iodo-2-fluoronitrobenzene
Formula: C₆H₂BrFINO₂
MW: 345.892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28096556
CAS Number: 1805552-21-9