2,2,3,3-Tetrafluoro-1,4-benzodioxane-6-carboxylic acid
Supplier: Apollo Scientific
Warning
PC500576-1G PC500576-250MG
APOSPC500576-1GEA 862 EUR
APOSPC500576-1G APOSPC500576-250MG
2,2,3,3-Tetrafluoro-1,4-benzodioxane-6-carboxylic acid
2,2,3,3-Tetrafluoro-1,4-benzodioxane-6-carboxylic acid
Formula: C₉H₄F₄O₄
MW: 252.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06208646
CAS Number: 180091-38-7