5-Bromo-4-(trifluoromethyl)isatoic anhydride
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromo-1,4-dihydro-2,4-dioxo-7-(trifluoromethyl)-2H-3,1-benzoxazine, 6-Bromo-7-(trifluoromethyl)-2H-3,1-benzoxazine-2,4(1H)-dione
PC500605-1G PC500605-250MG
APOSPC500605-1GEA 720 EUR
APOSPC500605-1G APOSPC500605-250MG
5-Bromo-4-(trifluoromethyl)isatoic anhydride
5-Bromo-4-(trifluoromethyl)isatoic anhydride
Formula: C₉H₃BrF₃NO₃
MW: 310.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103324
CAS Number: 1820740-28-0