3-Fluoro-4-(hydroxymethyl)benzonitrile 99+%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Cyano-2-fluorobenzyl alcohol
PC500665-1G PC500665-5G PC500665-250MG
APOSPC500665-1GEA 24.8 EUR
APOSPC500665-1G APOSPC500665-5G APOSPC500665-250MG
3-Fluoro-4-(hydroxymethyl)benzonitrile 99+%
3-Fluoro-4-(hydroxymethyl)benzonitrile
Formula: C₈H₆FNO
MW: 151.138 g/mol
Melting Pt: 63…68 °C
Storage Temperature: Ambient
MDL Number: MFCD09037779
CAS Number: 219873-06-0