2,4-Difluoro-3-nitropyridine
Supplier: Apollo Scientific
Warning
PC50084-250MG PC50084-1G
APOSPC50084-250MGEA 248 EUR
APOSPC50084-250MG APOSPC50084-1G
2,4-Difluoro-3-nitropyridine
2,4-Difluoro-3-nitropyridine
Formula: C₅H₂F₂N₂O₂
MW: 160.078 g/mol
Melting Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD18802742
CAS Number: 60186-14-3