2-(2,2-Difluoroethoxy)bromobenzene
Supplier: Apollo Scientific
Synonyms: 1-Bromo-2-(2,2-difluoroethoxy)benzene
PC500912-1G PC500912-5G
APOSPC500912-1GEA 161 EUR
APOSPC500912-1G APOSPC500912-5G
2-(2,2-Difluoroethoxy)bromobenzene
2-(2,2-Difluoroethoxy)bromobenzene
Formula: C₈H₇BrF₂O
MW: 237.041 g/mol
Boiling Pt: 133 °C (5 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11036116
CAS Number: 946605-55-6