3-Bromo-5-fluoro-4-iodobenzotrifluoride 95%
Supplier: Apollo Scientific
Warning
PC501118-1G PC501118-250MG PC501118-5G
APOSPC501118-1GEA 1070 EUR
APOSPC501118-1G APOSPC501118-250MG APOSPC501118-5G
3-Bromo-5-fluoro-4-iodobenzotrifluoride 95%
3-Bromo-5-fluoro-4-iodobenzotrifluoride
Formula: C₇H₂BrF₄I
MW: 368.893 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28348453
CAS Number: 1934724-90-9