4-Bromo-2,3-difluoro-6-nitroaniline
Supplier: Apollo Scientific
PC501256-1G PC501256-5G
APOSPC501256-1GEA 251 EUR
APOSPC501256-1G APOSPC501256-5G
4-Bromo-2,3-difluoro-6-nitroaniline
4-Bromo-2,3-difluoro-6-nitroaniline
Formula: C₆H₃BrF₂N₂O₂
MW: 253.001 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22375090
CAS Number: 626238-73-1