5-Bromo-3-fluoroisatoic anhydride
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromo-8-fluoro-2H-3,1-benzoxazine-2,4(1H)-dione
PC501400-1G PC501400-5G
APOSPC501400-1GEA 360 EUR
APOSPC501400-1G APOSPC501400-5G
5-Bromo-3-fluoroisatoic anhydride
5-Bromo-3-fluoroisatoic anhydride
Formula: C₈H₃BrFNO₃
MW: 260.017 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11899541
CAS Number: 1049118-00-4