4-Chloro-3-(trifluoromethoxy)nitrobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-4-nitro-2-(trifluoromethoxy)benzene, 2-Chloro-5-nitrophenyl trifluoromethyl ether
PC501403-1G PC501403-250MG PC501403-5G
APOSPC501403-1GEA 301 EUR
APOSPC501403-1G APOSPC501403-250MG APOSPC501403-5G
4-Chloro-3-(trifluoromethoxy)nitrobenzene
4-Chloro-3-(trifluoromethoxy)nitrobenzene
Formula: C₇H₃ClF₃NO₃
MW: 241.552 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393864
CAS Number: 113421-97-9