3,5-Difluoro-1-(trifluoromethoxy)benzene 99%
Supplier: Apollo Scientific
Danger
PC501509-1G PC501509-250MG PC501509-5G
APOSPC501509-1GEA 139 EUR
APOSPC501509-1G APOSPC501509-250MG APOSPC501509-5G
3,5-Difluoro-1-(trifluoromethoxy)benzene 99%
3,5-Difluoro-1-(trifluoromethoxy)benzene
Formula: C₇H₃F₅O
MW: 198.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22581043
CAS Number: 1404194-50-8
UN: 1993
ADR: 3,II