Methyl 3-fluoro-4-(hydroxymethyl)benzoate 97%
Supplier: Apollo Scientific
Warning
PC501575-1G PC501575-5G
APOSPC501575-1GEA 37.7 EUR
APOSPC501575-1G APOSPC501575-5G
Methyl 3-fluoro-4-(hydroxymethyl)benzoate 97%
Methyl 3-fluoro-4-(hydroxymethyl)benzoate
Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09836511
CAS Number: 937636-18-5