2-Bromo-4,6-difluorobenzaldehyde, tech
Supplier: Apollo Scientific
Warning
PC501584-1G PC501584-250MG PC501584-5G
APOSPC501584-1GEA 270 EUR
APOSPC501584-1G APOSPC501584-250MG APOSPC501584-5G
2-Bromo-4,6-difluorobenzaldehyde, tech
2-Bromo-4,6-difluorobenzaldehyde
Formula: C₇H₃BrF₂O
MW: 220.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18642940
CAS Number: 154650-59-6