3-Fluoro-5-nitrobenzenesulphonyl chloride 95%
Supplier: Apollo Scientific
Danger
PC501631-1G PC501631-250MG
APOSPC501631-1GEA 1640 EUR
APOSPC501631-1G APOSPC501631-250MG
3-Fluoro-5-nitrobenzenesulphonyl chloride 95%
3-Fluoro-5-nitrobenzenesulphonyl chloride
Formula: C₆H₃ClFNO₄S
MW: 239.609 g/mol
Boiling Pt: 136…138 °C
Melting Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD12197272
CAS Number: 1193388-16-7
UN: 3261
ADR: 8,III