Methyl 2-bromo-4,6-difluorobenzoate 97%
Supplier: Apollo Scientific
Warning
PC501649-1G PC501649-250MG
APOSPC501649-1GEA 248 EUR
APOSPC501649-1G APOSPC501649-250MG
Methyl 2-bromo-4,6-difluorobenzoate 97%
Methyl 2-bromo-4,6-difluorobenzoate
Formula: C₈H₅BrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18642939
CAS Number: 1379336-54-5