2-Bromo-4,6-difluorobenzyl alcohol
Supplier: Apollo Scientific
Warning
PC501657-1G PC501657-250MG
APOSPC501657-1GEA 343 EUR
APOSPC501657-1G APOSPC501657-250MG
2-Bromo-4,6-difluorobenzyl alcohol
2-Bromo-4,6-difluorobenzyl alcohol
Formula: C₇H₅BrF₂O
MW: 223.015 g/mol
Melting Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD28737914
CAS Number: 1807071-21-1