4-(Pentafluorothio)benzeneboronic acid, pinacol ester 95%
Supplier: Apollo Scientific
Warning
PC501675-1G PC501675-5G
APOSPC501675-1GEA 575 EUR
APOSPC501675-1G APOSPC501675-5G
4-(Pentafluorothio)benzeneboronic acid, pinacol ester 95%
4-(Pentafluorothio)benzeneboronic acid, pinacol ester
Formula: C₁₂H₁₆BF₅O₂S
Storage Temperature: Ambient
MDL Number: MFCD20489194
CAS Number: 1286278-34-9