2-(Difluoromethoxy)-5-fluoroaniline
Supplier: Apollo Scientific
Warning
PC501900-1G PC501900-250MG PC501900-5G
APOSPC501900-1GEA 347 EUR
APOSPC501900-1G APOSPC501900-250MG APOSPC501900-5G
2-(Difluoromethoxy)-5-fluoroaniline
2-(Difluoromethoxy)-5-fluoroaniline
Formula: C₇H₆F₃NO
MW: 177.124 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698442
CAS Number: 1214329-65-3