5-Bromo-2,3-difluorobenzoic acid
Supplier: Apollo Scientific
Warning
PC501969-1G PC501969-5G PC501969-250MG
APOSPC501969-1GEA 137 EUR
APOSPC501969-1G APOSPC501969-5G APOSPC501969-250MG
5-Bromo-2,3-difluorobenzoic acid
5-Bromo-2,3-difluorobenzoic acid
Formula: C₇H₃BrF₂O₂
MW: 236.998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972472
CAS Number: 887585-64-0