1-Bromo-1,1,2,2-tetrafluorohexane
Supplier: Apollo Scientific
Warning
PC50202-1G PC50202-5G PC50202-250MG
APOSPC50202-1GEA 0 EUR
APOSPC50202-1G APOSPC50202-5G APOSPC50202-250MG
1-Bromo-1,1,2,2-tetrafluorohexane
1-Bromo-1,1,2,2-tetrafluorohexane
Formula: C₆H₉BrF₄
MW: 237.033 g/mol
Boiling Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD25541897
CAS Number: 151831-45-7