Methyl 2-bromo-3-fluorobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-3-fluorobenzoic acid methyl ester
PC502130-1G PC502130-250MG PC502130-5G
APOSPC502130-1GEA 95.7 EUR
APOSPC502130-1G APOSPC502130-250MG APOSPC502130-5G
Methyl 2-bromo-3-fluorobenzoate
Methyl 2-bromo-3-fluorobenzoate
Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000921
CAS Number: 647020-71-1