3-Iodo-2-hydroxy-5-(trifluoromethoxy)benzaldehyde 95%
Supplier: Apollo Scientific
Warning
PC502222-1G PC502222-250MG PC502222-5G
APOSPC502222-1GEA 248 EUR
APOSPC502222-1G APOSPC502222-250MG APOSPC502222-5G
3-Iodo-2-hydroxy-5-(trifluoromethoxy)benzaldehyde 95%
3-Iodo-2-hydroxy-5-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄F₃IO₃
MW: 332.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25967134
CAS Number: 775330-11-5