5-Bromo-2-fluoro-3-(trifluoromethyl)aniline 97%
Supplier: Apollo Scientific
PC502237-1G PC502237-250MG PC502237-5G
APOSPC502237-1GEA 507 EUR
APOSPC502237-1G APOSPC502237-250MG APOSPC502237-5G
5-Bromo-2-fluoro-3-(trifluoromethyl)aniline 97%
5-Bromo-2-fluoro-3-(trifluoromethyl)aniline
Formula: C₇H₄BrF₄N
MW: 258.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28740802
CAS Number: 1805580-19-1